Butyl Rubber Injection Stopper, MicroSolv
Supplier: MicroSolv Technology Corp
95025-06
10014-072PK 82.84 USD
10014-072
Butyl Rubber Injection Stopper, MicroSolv
Stoppers Rubber Stoppers
Shaped, injection stoppers, butyl rubber used in PerkinElmer Injectors. Use in aluminum or steel crimp caps for headspace autosamplers which are sold separately.
Packaging: Supplied in polyethylene bags.