1,2-Dimyristoyl-sn-glycero-3-phosphocholine solution ≥99% in chloroform (14:0 PC)
Supplier: AVANTI POLAR LIPIDS INC MS
Synonyms:
14:0 PC, 1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(tetradecanoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, DMPC, β-γ-Dimyristoyl-L-α-Lecithin, (R)-2,3-Bis(tetradecanoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate
850345C25MG
850345C1G
850345C200MG
850345C500MG
100126-642EA
93.85
USD
100126-642
100121-646
100125-174
100122-994
1,2-Dimyristoyl-sn-glycero-3-phosphocholine solution ≥99% in chloroform (14:0 PC)
1,2-Dimyristoyl-sn-glycero-3-phosphocholine
- Purified by HPLC
- Protected from oxidization and hydrolysis
Formula:
C₃₆H₇₂NO₈P MW: 677.94 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00036901 CAS Number: 18194-24-6 |
Specification Test Results
Purity | >99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...