Axygen® PCR Tubes and Caps, Corning
Supplier: Corning
PCR-02-A PCR-02-C PCR-02D-A PCR-02D-C PCR-02-NC PCR-05-A PCR-05-C PCR-02-C-500 PCR-05-C-500
10011-776CS 1364.98 USD
10011-776 10011-780 10011-798 10011-802 10011-816 10011-826 10011-830 10169-888 10169-890
Axygen® PCR Tubes and Caps, Corning
Tubes PCR Tubes
Axygen thin wall PCR strip tubes are designed to fit a variety of thermal cyclers. The consistent and ultra-thin wall allows precise thermal transfer.
An even insertion cap ensures a tight fit to reduce evaporation. Strip tubes are available in an assortment of colors.
Optical flat cap strips feature ultra-clear upper surfaces, ideal for fluorescent luminescent and real-time PCR applications.
They offer a light transmittance that is 75% higher than with standard strip caps. Made to seal tube strips and 96-well microplates.