Axygen® Snaplock Microcentrifuge Tubes, 0.6/1.5/1.7/2.0/5.0 ml, Corning
Supplier: Corning
MCT-175-SP MCT-200-SP MCT-060-B MCT-060-C MCT-060-C-S MCT-060-G MCT-060-O MCT-060-R MCT-060-V MCT-060-X MCT-150-A MCT-150-B MCT-150-C MCT-150-C-S MCT-150-G MCT-150-O MCT-150-R MCT-150-V MCT-150-X MCT-150-Y MCT-175-A MCT-175-B MCT-175-C MCT-175-C-S MCT-175-G MCT-175-O MCT-175-R MCT-175-V MCT-175-X MCT-175-Y MCT-200-A MCT-200-B MCT-200-C MCT-200-C-S MCT-200-G MCT-200-O MCT-200-R MCT-200-V MCT-200-X MCT-060-L-C MCT-150-L-C MCT-175-L-C MCT-200-L-C MCT-500-C MCT-500-C-S MCT-500-X
80086-026CS 671.42 USD
80086-026 80086-028 10011-678 10011-680 10011-682 10011-684 10011-686 10011-688 10011-690 10011-692 10011-694 10011-696 10011-698 10011-700 10011-702 10011-704 10011-708 10011-710 10011-712 10011-714 10011-716 10011-718 10011-720 10011-722 10011-724 10011-726 10011-728 10011-730 10011-732 10011-734 10011-736 10011-738 10011-740 10011-742 10011-744 10011-746 10011-750 10011-752 10011-754 10011-756 22234-042 22234-044 22234-046 22234-048 89425-666 89425-665 89425-667 89425-669
Axygen® Snaplock Microcentrifuge Tubes, 0.6/1.5/1.7/2.0/5.0 ml, Corning
Tubes Microcentrifuge Tubes
High quality PP, graduated.
  • Maxymum Recovery® microtubes with ultra-smooth surfaces prevent adhesion of proteins and nucleic acids after long periods of storage
  • Certified free from DNase, RNase and endotoxin
Microtubes made from PP homopolymer for excellent sample visibility. Available in an ultra-transparent (MaxyClear) version clear or in assorted colours, with frosted writing area.