Orion™ Chlorine Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
9770BNWP
10010-830EA 875.76 USD
10010-830
Orion™ Chlorine Electrodes, Thermo Scientific
Electrodes ISE Electrodes
Measure total residual chlorine quickly and accurately with Orion™ Chlorine Electrodes
All forms of total residual chlorine are measured.