Orion™ Cadmium Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
9648BNWP
10010-818EA 1165.9 USD
10010-818
Orion™ Cadmium Electrodes, Thermo Scientific
Electrodes ISE Electrodes
Measure Cadmium ions in aqueous solutions quickly, simply, and economically with Orion™ Cadmium Electrodes.
  • Free-flowing Sure-Flow junction assures stable, drift-free potentials
  • A separate reference electrode is unnecessary
  • Convenient for use with small sample volumes