Orion™ Cyanide Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
9606BNWP
34116-968EA 1982.46 USD
34116-968 10010-806
Orion™ Cyanide Electrodes, Thermo Scientific
Electrodes ISE Electrodes
Measure cyanide ions in aqueous solutions quickly, simply, and economically with Orion™ Cyanide Electrodes.
  • Free-flowing Sure-Flow junction assures stable, drift-free potentials
  • A separate reference electrode is unnecessary
  • Convenient for use with small sample volumes