Orion™ Ammonia Ion Selective Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
9512BNWP
10010-804EA 978 USD
10010-804
Orion™ Ammonia Ion Selective Electrodes, Thermo Scientific
Electrodes ISE Electrodes
This standard gas-sensing ISE electrode is designed for quick, reliable and accurate measurements of dissolved ammonia in aqueous solutions.
  • Waterproof BNC connector
  • Can be used on any ISE or mV meter with a BNC connection
The ammonia combination electrode has the sensing and reference half-cells built into one electrode, which decreases the amount of required solutions and reduces waste.
Accessories information: Use Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes. Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes. Electrode handles are used with plastic membrane combination or half-cell electrodes.
Ordering information: Electrode is supplied with 40 loose membranes, one reusable membrane cap, tweezers for handling the membranes and one bottle of electrode filling solution with dispensing cap.