Orion™ Star™ Series RS232 to USB Cable Adapter, Thermo Scientific
Supplier: Thermo Fisher Scientific
1010005
10010-666EA 143.29 USD
10010-666
Orion™ Star™ Series RS232 to USB Cable Adapter, Thermo Scientific
Cables
This cable adapter can be used to connect a USB port to the RS232 port of a meter.
The adapter allows interface between a USB connection and Orion™ Versa Star™ meters, Orion™ Star A series (Star A320, Star A220 and Star A210 series) meters, and legacy Orion™ Star™ and Star Plus meters.