Orion™ Star™ Series RS232 Printer and Computer Cable Set, Thermo Scientific
Supplier: Thermo Fisher Scientific
1010004
10010-664EA 81.88 USD
10010-664
Orion™ Star™ Series RS232 Printer and Computer Cable Set, Thermo Scientific
Cables
This cable set can be used to connect a computer and printer to a benchtop or portable meter.
Designed for use with Orion™ Versa Star™ meters, Orion™ Star™ A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus meters.