Orion™ Star™ Series Soft-Sided Field Case for Portable Meters, Thermo Scientific
Supplier: Thermo Fisher Scientific
1210005
10010-474EA 175.33 USD
10010-474
Orion™ Star™ Series Soft-Sided Field Case for Portable Meters, Thermo Scientific
Electrode Holders
This cushioned field case with interior dividers and exterior pockets protects meters, electrodes, solution, and beakers under field conditions.
Designed for use with Orion™ Star™ A120 series portable meters; Orion™ Star™ A220 series portable meters; Orion™ Star™ A320 series portable meters; Orion™ 3-Star, 4-Star and 5-Star series portable meters; Orion™ Star™ and Star Plus portable meters.