Hard-Shell® 96-Well Semi-Skirted PCR Plates, Low-Profile, Bio-Rad

Supplier: Bio-Rad
HSL9905
100069-374EA 130 USD
100069-374 100069-376 100720-952 100720-956
Hard-Shell® 96-Well Semi-Skirted PCR Plates, Low-Profile, Bio-Rad
Microplates PCR Plates
Hard-Shell® 96-Well Semi-Skirted PCR Plates are designed to withstand the stresses of thermal cycling and robotic handling

  • Reaction volumes of 5–125 µl (200 µl maximum)
  • Black lettering for easy well identification
  • Free of DNase, RNase, and human DNA
  • White-well option for increased fluorescent signal strength
  • Color-coded skirt option with clear wells
  • Low-cost, user-readable barcode option for convenient database tracking

The half-height skirt provides improved robotic handling and a labeling surface, and the low-profile wells are optimized for low-volume reactions and fast PCR.

The patented two-component design provides stability and flatness, allowing precise positioning for automation. The skirt and deck are molded from a rigid thermostable polymer to eliminate warping and shrinkage, and the thin-wall wells are molded of virgin polypropylene with low DNA binding.

Compatible with: Bio-Rad's C1000™, C1000 Touch™, S1000™, CFX96™, CFX96 Touch™, CFX96 Touch™ Deep Well, CFX Connect™; Applied Biosystems 9800 fast, Veriti fast, 7500 fast, ViiA 7 fast; Eppendorf Mastercycler series, Eppendorf Mastercycler ep realplex.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR