BAX® System X5 PCR Assay for Escherichia coli O157:H7, Hygiena™, Qualicon Diagnostics LLC
Supplier: Hygiena
KIT2022
10002-884EA 708.6 USD
10002-884
BAX® System X5 PCR Assay for Escherichia coli O157:H7, Hygiena™, Qualicon Diagnostics LLC
Microbiology Test Systems PCR Tests
The BAX® System X5 PCR Assay for E
Each BAX System X5 PCR assay includes all the reagents you need to process 64 enriched samples – enough for two full process runs in the BAX System X5 instrument. BAX System X5 PCR tablets contain a built-in control and are pre-packaged in PCR tubes to eliminate the need for reagent preparation.
The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.
coli O157:H7 detects the target organism from standard enrichments of a variety of food and environmental samples. The method includes automated sample preparation in the DuPont™ thermal block, followed by automated amplification and detection in the BAX System X5 instrument.
Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System X5 instrument.