BAX® System X5 Start-Up Package, Hygiena™, Qualicon Diagnostics LLC
Supplier: Hygiena
MCH2007
10002-882EA 25175.09 USD
10002-882
BAX® System X5 Start-Up Package, Hygiena™, Qualicon Diagnostics LLC
Microbiology Test Systems PCR Tests
The Hygiena™ BAX® System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect foodborne pathogens, spoilage organisms and other microbes in raw ingredients, finished products and environmental samples.
The BAX® System X5 offers the same fast, accurate and easy-to-use pathogen detection solution that customers have come to expect from the BAX® System method, but in a smaller lightweight construction to provide greater testing flexibility
After a standard enrichment procedure, automated sample preparation in the Hygiena™ Thermal Block is complete in about 1 hour, and automated PCR processing is complete in the BAX® System X5 instrument in 3.5 hours.
BAX® System X5 instrument is simple to install and virtually maintenance-free, and handles up to 32 samples at a time to best fit smaller testing volume requirements and bench space limitations.
Ordering information: The Hygiena™ BAX® System X5 Startup Package includes one BAX® System X5 instrument with power cables and network cable; laptop computer pre-loaded with BAX® System X5 software; Hygiena™ Thermal Block with metal tube holders; 32-well aluminum cold block with insulator for PCR tubes; two 32-well sample holders; two BAX® System capping/de-capping tools; multi-channel, adjustable, and repeating pipettes with tips; cluster tubes with caps and racks; PCR tube caps; powder free nitrile gloves; user documentation on USB memory drive.