BAX® System X5 PCR Assay for Listeria, Hygiena™, Qualicon Diagnostics LLC
Supplier: Hygiena
KIT2024
10002-772EA 708.6 USD
10002-772
BAX® System X5 PCR Assay for Listeria, Hygiena™, Qualicon Diagnostics LLC
Microbiology Test Systems PCR Tests
The BAX® System X5 PCR Assay for genus Listeria detects the target organism from standard enrichments of a variety of food and environmental samples
Each BAX System X5 PCR assay includes all the reagents you need to process 64 enriched samples – enough for two full process runs in the BAX® System X5 instrument. BAX System X5 PCR tablets contain a built-in control and are pre-packaged in PCR tubes to eliminate the need for reagent preparation.
The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.
The method includes automated sample preparation in the DuPont™ thermal block, followed by automated amplification and detection in the BAX® System X5 instrument.
Ordering information: Includes PCR tubes with tablets, optical caps, Lysing Agent 2, protease, and lysis buffer. This assay can only be used with the BAX System X5 instrument.