VWR® Freezer Box for Centrifuge Tubes
Supplier: VWR International
10002-734
10002-734PK 118.12 USD
10002-734
VWR® Freezer Box for Centrifuge Tubes
Boxes Cryoboxes
VWR® Freezer Box for Centrifuge Tubes features polycarbonate construction and is supplied with lid.
  • Suitable over a range of temperatures
  • Polycarbonate construction
The box is suitable for use in temperatures as low as -190°C (-310°F). Each box will accommodate 25 x 5 mL centrifuge tubes.