HyFlex® 11-724 Medium Duty, Cut Resistant Gloves, Ansell
Supplier: Ansell Healthcare
11724100
10002-056CS 2051.67 USD
10002-056
HyFlex® 11-724 Medium Duty, Cut Resistant Gloves, Ansell
Gloves
Excellent flexibility with optimal cut protection, the HyFlex® 11-724 glove features a 13G lightweight, medium-duty liner with INTERCEPT TECHNOLOGY® yarns that provide EN Cut Level 3 (ANSI Level 2) protection.
  • Silicone-free
  • Increased worker confidence when handling small, sharp parts
  • Exceptional abrasion resistance
  • Minimal shrinkage after laundering
The thin, ergonomic liner features an optimized Lycra® content that is soft and cool on the skin for all-day comfort
Precision, dexterity, and flexibility when handling small, sharp parts increases worker confidence while on the job. Durable and launderable with minimal shrinkage and excellent recovery after laundering, extending glove life.
INTERCEPT TECHNOLOGY® yarn has been tested to perform at or exceed industry standards. This Ansell proprietary technology offers cut protection and increased comfort and dexterity. The fully coated design improves worker grip, extends glove life in abrasive conditions and allows the back of hand to breathe.
For use in aerospace, automotive, machinery and equipment and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light-duty stamping and maintenance.
Certifications: ANSI Cut Level 2. ANSI Abrasion Level 6.
Packaging: Packaged 12 pairs in a bag; 12 bags in a carton.