Ultra Bright Blue LED Transilluminator, G-Biosciences
Supplier: G-Biosciences
BT507
10001-872EA 2188.44 USD
10001-872
Ultra Bright Blue LED Transilluminator, G-Biosciences
Transilluminators Blue Light Transilluminators
The Ultra Bright Blue LED Transilluminator has a strong blue LED light for exitation that does not cause durn to your eyes or skin.
For use with nucleic acid and protein stains such LabSafe™ Nucleic Acid Stain, EtBr, SYBR® Safe, SYBR Gold, SYBR® Green I & II, SYPRO® Ruby, SYPRO® Orange, Coomassie Fluor™ Orange stains, GelGreen, GelRed and Lumitein™ Protein Gel Stain.
​Safer to replace UV, the Ultra Bright Blue LED transilluminator makes gel cutting easy - No need to wear amber glasses thanks to the adjustable intensity from 100% down to 50%. The strong blue LED for excitation doesMore Product Information