Mini Horizontal Gel Electrophoresis System, Geno-Electrophore, G-Biosciences
Supplier: G-Biosciences
BT105
10001-836EA 732.18 USD
10001-836
Mini Horizontal Gel Electrophoresis System, Geno-Electrophore, G-Biosciences
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
For a variety of agarose electrophoresis applications, the ultimate in simplicity and convenience in casting agarose gels
The casting trays are provided with background black strips for high well visibility. The combs provide either 13 and 6 wells (0.6 x 0.1 cm well) or 17 and 8 wells (0.4 x 0.1 cm well) for the large and mini gels, respectively. Interlocked ventilated fog free lid allows full view of gel during electrophoresis. Replacement parts are also available including additional combs (MT-C01, MT-C02), gel trays (MT-T01, MT-T02), casting tray (MT-T03) or a complete Gel Casting Set (1 Casting Tray, 2 Combs, 1 Large Tray & 2 Small Trays) (MT-T04).
Less agarose consumption leads to less running buffer consumption. Multichannel pipette-compatible. Single molded tank. Safety and ventilation lid enables to cast three pieces of agarose gel in one time. No direct heat impact from power supply unit (high temperature capability, 130°C), easy sample loading. Great indications for gel making and running. No tapes, clamps or springs needed for gel casting.
Light-weight and simple to use, the gel casting tray is designed to eliminate the leak problems common with other gel casting trays.
Certifications: Black well visualization strip and migration distance index line
Ordering information: Supplied with agarose gel casting trays, horizontal tank and two different size combs.