HiTrap Columns, AVB Sepharose HP
Supplier: Cytiva
28-4112-11 28-4112-12
10000-098EA 1647.71 USD
10000-098 10000-100
HiTrap Columns, AVB Sepharose HP
Chromatography Columns
AVB Sepharose High Performance is an affinity resin designed for the purification of adeno associated virus (AAV), Prepacked HiTrap columns for simple operation with a syringe, pump, or chromatographic system.
  • One step purification of adeno associated viruses
  • Affinity resin with high selectivity and low background binding
  • Large surface area to give enhanced capacity
AVB Sepharose High performance is an affinity resin with affinity for adeno associated viruses. It enables purification of adeno associated viruses in one step with high purity and excellent yields. It has proven affinity for adeno associated viruses from subclasses 1, 2, 3, and 5, but it is expected to have affinity for most of the other adeno associated virus subclasses. The ligand is a 14 kD fragment from a single chain antibody, expressed in yeast.